α,α-Difluoroanisole ≥99%
Supplier: Matrix Scientific
Synonyms:
(Difluoromethoxy)benzene, alpha,alpha-Difluoroanisole
004455-100G
004455-25G
101777-582EA
464.62
USD
101777-582
101777-580
α,α-Difluoroanisole ≥99%
α,α-Difluoroanisole
Hazard: Flammable
Formula:
C₇H₆F₂O MW: 144.1200 g/mol Boiling Pt: 98…100 °C Density: 1.2 Flash Pt: 38 °C |
MDL Number:
MFCD00068184 CAS Number: 458-92-4 |
Specification Test Results
Boiling point | 98-100° |
Density | 1.2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...