5-Fluoro-m-xylene ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5-Dimethylfluorobenzene, 1-Fluoro-3,5-dimethylbenzene, 5-Fluoro-1,3-dimethylbenzene

004442-5G 004442-25G
101777-566EA 49.23 USD
101777-566 101777-568
5-Fluoro-m-xylene ≥98%
5-Fluoro-m-xylene

Hazard: Flammable

Formula: C₈H₉F
MW: 124.16 g/mol
Boiling Pt: 145°C
Density: 0.98
MDL Number: MFCD00052366
CAS Number: 461-97-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 145°
Density 0.98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR