Diethyl fluoromalonate ≥97%

Supplier: Matrix Scientific

Synonyms: 1,3-diethyl 2-fluoropropanedioate

004404-25G 004404-100G
101777-550EA 133.85 USD
101777-550 102663-556
Diethyl fluoromalonate ≥97%
Diethyl fluoromalonate

Hazard: Toxic/Corrosive

Formula: C₇H₁₁FO₄
MW: 178.16 g/mol
Boiling Pt: 121…122 °C (30 mmHg)
Density: 1.129
MDL Number: MFCD00009139
CAS Number: 685-88-1

Order Now

Specification Test Results

Boiling point 121-122°/3
Density 1.129
ND20 1.407

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR