2-Chloro-2,2-dfluoroacetyl chloride ≥97%

Supplier: Matrix Scientific

004392-5G 004392-25G
101777-538EA 144.62 USD
101777-538 101777-540
2-Chloro-2,2-dfluoroacetyl chloride ≥97%
2-Chloro-2,2-dfluoroacetyl chloride

TCA listed: T
Hazard: Corrosive/Refrigerate

Formula: C₂Cl₂F₂O
Boiling Pt: 25°C
MDL Number: MFCD00039317
CAS Number: 354-24-5

Order Now

Specification Test Results

Boiling point 49-50°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR