Chlorodifluoroacetic anhydride ≥98%
Supplier: Matrix Scientific
004382-25G
004382-5G
101777-536EA
284.62
USD
101777-536
101777-534
Chlorodifluoroacetic anhydride ≥98%
Chlorodifluoroacetic anhydride
TCA listed: T
Hazard: Corrosive/Lachrymator
Formula:
C₄Cl₂F₄O₃ MW: 242.94 g/mol Boiling Pt: 96…97 °C Density: 1.395 |
MDL Number:
MFCD00000774 CAS Number: 2834-23-3 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 96-97° |
Density | 1.395 |
ND20 | 1.347 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...