Benzoyl fluoride ≥99%

Supplier: Matrix Scientific

004368-100G 004368-25G
101777-528EA 638.46 USD
101777-528 101777-526
Benzoyl fluoride ≥99%
Benzoyl fluoride

TCA listed: T
Hazard: Flammable/Corrosive/Lachrymator

Formula: C₇H₅FO
Boiling Pt: 159…161 °C
Melting Pt: –28 °C
Density: 1.14
MDL Number: MFCD00000364
CAS Number: 455-32-3
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Boiling point 159-161°
Density 1.14
ND20 1.496

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR