Trimethylsilylacetylene ≥98%

Supplier: Matrix Scientific

004348-25G 004348-100G
101777-510EA 84.62 USD
101777-510 101777-512
Trimethylsilylacetylene ≥98%
Trimethylsilylacetylene

TCA listed: T
Hazard: Flammable/Refrigerate

Formula: C₅H₁₀Si
Boiling Pt: 53°C
Density: 0.71
Flash Pt: -34°C
MDL Number: MFCD00008569
CAS Number: 1066-54-2

Order Now

Specification Test Results

Boiling point 53°
Density 0.695
ND20 1.388


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR