1-Naphthalenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

004304-5G 004304-25G
101777-486EA 73.85 USD
101777-486 101777-488
1-Naphthalenesulfonyl chloride ≥97%
1-Naphthalenesulphonyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₁₀H₇ClO₂S
Boiling Pt: 194…195 °C (13 mmHg)
Melting Pt: 66…68 °C
MDL Number: MFCD00003984
CAS Number: 85-46-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 194-195°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR