4-Propylbenzenesulphonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Propylbenzene-1-sulphonyl chloride

004287-5G 004287-25G
101777-478EA 187.69 USD
101777-478 101777-480
4-Propylbenzenesulphonyl chloride ≥95%
4-Propylbenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₉H₁₁ClO₂S
Boiling Pt: 275-276°C
MDL Number: MFCD00041508
CAS Number: 63014-04-0

Order Now

Specification Test Results

Boiling point 275-276°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR