2,2,4,4-Tetramethyl-3-pentanone ≥98%

Supplier: Matrix Scientific

Synonyms: Di-tert-butyl ketone, Hexamethylacetone

004268-5G 004268-25G
101777-466EA 93.85 USD
101777-466 101777-468
2,2,4,4-Tetramethyl-3-pentanone ≥98%
2,2,4,4-Tetramethyl-3-pentanone

TCA listed: T
Hazard: Flammable

Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 152-153°C
Density: 0.824
Flash Pt: 32°C
MDL Number: MFCD00008845
CAS Number: 815-24-7
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Boiling point 152-153°
Density 0.824
ND20 1.419

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR