1-Iodo-7-(trifluoromethyl)perfluorooctane ≥98%

Supplier: Matrix Scientific

004236-5G 004236-25G
101777-450EA 50.77 USD
101777-450 101777-452
1-Iodo-7-(trifluoromethyl)perfluorooctane ≥98%
1-Iodo-7-(trifluoromethyl)perfluorooctane

TCA listed: T
Hazard: Irritant

Formula: C₉F₁₉I
MW: 595.97 g/mol
Boiling Pt: 190°C
MDL Number: MFCD00153243
CAS Number: 865-77-0

Order Now

Specification Test Results

Boiling point 190°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR