5-Bromo-2-thiophenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

004205-5G 004205-25G 004205-100G
101777-430EA 18.46 USD
101777-430 101777-432 101777-434
5-Bromo-2-thiophenesulfonyl chloride ≥95%
5-Bromo-2-thiophenesulphonyl chloride

Hazard: Corrosive/Irritant-Harmful/Moisture Sensitive

Formula: C₄H₂BrClO₂S₂
Boiling Pt: 99-101°C/0.5mm
Melting Pt: 38-40°C
MDL Number: MFCD00084909
CAS Number: 55854-46-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 99-101°/0.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR