3-Bromothiophene ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Thienyl bromide
004199-100G
004199-500G
101777-426EA
100
USD
101777-426
101777-428
3-Bromothiophene ≥97%
3-Bromothiophene
TCA listed: T
Hazard: Flammable/Toxic
Formula:
C₄H₃BrS MW: 163.04 g/mol Boiling Pt: 150°C Melting Pt: Density: 1.74 Flash Pt: 56°C |
MDL Number:
MFCD00005464 CAS Number: 872-31-1 UN: 2810 ADR: 6.1,II |
Specification Test Results
Boiling point | 150° |
Density | 1.74 |
ND20 | 1.591 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...