Methyl trifluoropyruvate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate

004156-5G 004156-100G 004156-25G
101777-406EA 18.46 USD
101777-406 101777-410 101777-408
Methyl trifluoropyruvate ≥97%
Methyl trifluoropyruvate

Hazard: Flammable/Irritant

Formula: C₄H₃F₃O₃
MW: 156.06 g/mol
Boiling Pt: 84…86 °C
Density: 1.528
Flash Pt: 26 °C
MDL Number: MFCD00114936
CAS Number: 13089-11-7

Order Now

Specification Test Results

Boiling point 84-86°
Density 1.528
ND20 1.332

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR