(3,3,3-Trifluoropropyl)trichlorosilane ≥97%

Supplier: Matrix Scientific

004151-25G 004151-100G
101777-402EA 87.69 USD
101777-402 101777-404
(3,3,3-Trifluoropropyl)trichlorosilane ≥97%
(3,3,3-Trifluoropropyl)trichlorosilane

TCA listed: T
Hazard: Flammable/Corrosive/Refrigerate

Formula: F₃CCH₂CH₂SiCl₃
Boiling Pt: 113…114 °C
Density: 1.395
Flash Pt: 15 °C
MDL Number: MFCD00013602
CAS Number: 592-09-6
UN: 2985
ADR: 3,II

Order Now

Specification Test Results

Boiling point 113-114°
Density 1.395
ND20 1.385


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR