Phenyl trifluoromethyl sulfide ≥98%

Supplier: Matrix Scientific

Synonyms: Phenyl(trifluoromethyl)sulphane, alpha,alpha,alpha-Trifluorothioanisole, (Trifluoromethylthio)benzene

004146-5G 004146-1G 004146-25G
101777-396EA 50.77 USD
101777-396 101777-394 101777-398
Phenyl trifluoromethyl sulfide ≥98%
Phenyl trifluoromethyl sulfide

Hazard: Flammable/Stench

Formula: C₇H₅F₃S
Boiling Pt: 140-142°C
Density: 1.249
Flash Pt: 40°C
MDL Number: MFCD00040839
CAS Number: 456-56-4

Order Now

Specification Test Results

Boiling point 140-142°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR