Methyl-4,4,4-trifluoroacetoacetate ≥95%
Supplier: Matrix Scientific
004126-5G
004126-25G
004126-100G
101777-376EA
161.54
USD
101777-376
101777-378
101777-380
Methyl-4,4,4-trifluoroacetoacetate ≥95%
Methyl-4,4,4-trifluoroacetoacetate
Hazard: Flammable
Formula:
C₅H₅F₃O₃ MW: 170.09 Boiling Pt: 117-118°C Density: 1.34 |
MDL Number:
MFCD00041004 CAS Number: 83643-84-9 UN: 3272 ADR: 3,III |
Specification Test Results
Boiling point | 117-118° |
Density | 1.34 |
ND20 | 1.3721 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...