2-Chloronicotinonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-cyanopyridine, 2-Chloropyridine-3-carbonitrile

004113-25G 004113-100G
101777-368EA 49.23 USD
101777-368 101777-370
2-Chloronicotinonitrile ≥98%
2-Chloronicotinonitrile

Hazard: Toxic/Irritant-Harmful

Formula: C₆H₃ClN₂
MW: 138.56 g/mol
Boiling Pt: 112 °C (1 mmHg)
Melting Pt: 106…109 °C
MDL Number: MFCD00014628
CAS Number: 6602-54-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 112°/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR