4-Pentenoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Allylacetic acid, Pent-4-enoic acid

004109-5G 004109-25G 004109-100G
101777-362EA 21.54 USD
101777-362 101777-364 101777-366
4-Pentenoic acid ≥97%
4-Pentenoic acid

TCA listed: T
Hazard: Toxic/Corrosive

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 83…84 °C (12 mmHg)
Melting Pt: –22.5 °C
Density: 0.981
MDL Number: MFCD00004408
CAS Number: 591-80-0
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Boiling point 83-84°/12m
Density 0.981
ND20 1.4283

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR