2,5-Dimethylthiophene ≥98%

Supplier: Matrix Scientific

004103-100G 004103-25G
101777-360EA 269.23 USD
101777-360 101777-358
2,5-Dimethylthiophene ≥98%
2,5-Dimethylthiophene

Hazard: Flammable/Stench

Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 134 °C (740 mmHg)
Density: 0.985
MDL Number: MFCD00005452
CAS Number: 638-02-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 134°/740mm
Density 0.985
ND20 1.512

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR