1-(Trimethylsilyl)imidazole ≥97%
Supplier: Matrix Scientific
004094-25G
004094-100G
101777-354EA
38.46
USD
101777-354
101777-356
1-(Trimethylsilyl)imidazole ≥97%
1-(Trimethylsilyl)imidazole
TCA listed: T
Hazard: Flammable/Moisture Sensitive/Refrigerate
Formula:
C₆H₁₂N₂Si Boiling Pt: 93…94 °C (14 mmHg) Density: 0.956 |
MDL Number:
MFCD00005280 CAS Number: 18156-74-6 |
Specification Test Results
Boiling point | 93-94°/14m |
Density | 0.956 |
ND20 | 1.475 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...