3,4-Xylyl isothiocyanate ≥99%

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethylphenyl isothiocyanate

004087-1G 004087-5G
101777-346EA 15.38 USD
101777-346 101777-348
3,4-Xylyl isothiocyanate ≥99%
3,4-Xylyl isothiocyanate

Hazard: Toxic/Corrosive

Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 132 °C (8 mmHg)
Density: 1.08
MDL Number: MFCD00031536
CAS Number: 19241-17-9

Order Now

Specification Test Results

Boiling point 132°/8mm
Density 1.08

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR