2,3-Xylyl isothiocyanate ≥97%

Supplier: Matrix Scientific

Synonyms: 2,3-Dimethylphenyl isothiocyanate

004086-5G 004086-1G
101777-344EA 198.46 USD
101777-344 101777-342
2,3-Xylyl isothiocyanate ≥97%
2,3-Xylyl isothiocyanate

Hazard: Toxic/Corrosive

Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 128°C/8mm
Density: 1.08
MDL Number: MFCD00041064
CAS Number: 1539-20-4

Order Now

Specification Test Results

Boiling point 128°/8mm
Density 1.08

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR