2,5-Xylyl isothiocyanate ≥98%
Supplier: Matrix Scientific
Synonyms:
2,5-Dimethylphenylisothiocyanate, 2,5-Dimethylphenyl isothiocyanate
004084-5G
004084-25G
101777-334EA
56.92
USD
101777-334
101777-336
2,5-Xylyl isothiocyanate ≥98%
2,5-Xylyl isothiocyanate
Hazard: Toxic/Corrosive
Formula:
C₉H₉NS MW: 163.24 g/mol Boiling Pt: 120 °C (8 mmHg) Density: 1.08 |
MDL Number:
MFCD00046798 CAS Number: 19241-15-7 |
Specification Test Results
Boiling point | 120°/8mm |
Density | 1.08 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...