4-Fluorophthalic anhydride ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Fluoroisobenzofuran-1,3-dione

004031-5G 004031-25G
101777-302EA 146.15 USD
101777-302 101777-304
4-Fluorophthalic anhydride ≥97%
4-Fluorophthalic anhydride

TCA listed: T
Hazard: Toxic/Moisture Sensitive

Formula: C₈H₃FO₃
Melting Pt: 79 °C
MDL Number: MFCD00191363
CAS Number: 319-03-9

Order Now

Specification Test Results

Melting point 79°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR