3,4-Dimethoxybenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethoxybenzene-1-sulphonyl chloride

003995-25G 003995-5G
101777-290EA 93.85 USD
101777-290 101777-288
3,4-Dimethoxybenzenesulfonyl chloride ≥98%
3,4-Dimethoxybenzenesulphonyl chloride

Hazard: Corrosive/Irritant-Harmful/Moisture Sensitive

Formula: C₈H₉ClO₄S
MW: 236.68 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD00051769
CAS Number: 23095-31-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 64-66°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR