2,5-Dimethoxybenzenesulphonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Dimethoxybenzene-1-sulphonyl chloride

003994-5G 003994-25G
101777-284EA 30.77 USD
101777-284 101777-286
2,5-Dimethoxybenzenesulphonyl chloride ≥98%
2,5-Dimethoxybenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₈H₉ClO₄S
MW: 236.68
Melting Pt: 109…111 °C
MDL Number: MFCD00024872
CAS Number: 1483-28-9
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Melting point 109-111°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR