3-Chlorothiophenol ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Chlorobenzene-1-thiol, 1-Chloro-3-mercaptobenzene, 3-Chlorobenzenethiol, m-Chlorothiophenol
003970-25G
003970-100G
101777-272EA
106.15
USD
101777-272
101777-274
3-Chlorothiophenol ≥97%
3-Chlorothiophenol
TCA listed: T
Hazard: Corrosive/Stench
Formula:
C₆H₅ClS MW: 144.62 g/mol Boiling Pt: 110 °C (30 mmHg) Density: 1.245 |
MDL Number:
MFCD00004839 CAS Number: 2037-31-2 |
Specification Test Results
Boiling point | 110°/30mm |
Density | 1.245 |
ND20 | 1.6021 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...