2-Methylthiophene ≥98%

Supplier: Matrix Scientific

003958-100G 003958-25G
101777-258EA 98.46 USD
101777-258 101777-256
2-Methylthiophene ≥98%
2-Methylthiophene

TCA listed: T
Hazard: Flammable/Stench

Formula: C₅H₆S
MW: 98.17 g/mol
Boiling Pt: 113°C
Density: 1.014
MDL Number: MFCD00005451
CAS Number: 554-14-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 113°
Density 1.014
ND20 1.52

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR