Undecafluorohexanoic acid ≥98%

Supplier: Matrix Scientific

003945-5G 003945-25G
101777-248EA 73.85 USD
101777-248 101777-250
Undecafluorohexanoic acid ≥98%
Undecafluorohexanoic acid

TCA listed: T
Hazard: Corrosive

Formula: C₆HF₁₁O₂
Boiling Pt: 157 °C
Density: 1.762
MDL Number: MFCD00198040
CAS Number: 307-24-4

Order Now

Specification Test Results

Boiling point 157°
Density 1.762
ND20 1.298


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR