(4-Fluorophenyl)acetyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluorobenzeneacetyl chloride, 4-Fluorophenylacetyl Chloride

003906-25G 003906-100G
101777-230EA 61.54 USD
101777-230 101777-232
(4-Fluorophenyl)acetyl chloride ≥97%
(4-Fluorophenyl)acetyl chloride

Hazard: Corrosive/Irritant/Moisture Sensitive

Formula: (C₆H₄F)CH₂COCl
MW: 172.59 g/mol
Boiling Pt: 60°C/8mm
Density: 1.259
MDL Number: MFCD00674181
CAS Number: 459-04-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 60°/8mm
Density 1.259
ND20 1.512

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR