(3-Fluorophenyl)acetonitrile ≥99%

Supplier: Matrix Scientific

Synonyms: 2-(3-Fluorophenyl)acetonitrile, 3-Fluorobenzyl cyanide

003900-100G 003900-25G
101777-220EA 120 USD
101777-220 101777-218
(3-Fluorophenyl)acetonitrile ≥99%
(3-Fluorophenyl)acetonitrile

TCA listed: T
Hazard: Toxic/Irritant

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 113…114 °C (18 mmHg)
Density: 1.163
MDL Number: MFCD00001907
CAS Number: 501-00-8
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 113-114°/1
Density 1.163
ND20 1.502

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR