2-Phenylpropionitrile ≥97%

Supplier: Matrix Scientific

Synonyms: alpha-Methylphenylacetonitrile

003893-5G 003893-25G
101777-210EA 30.77 USD
101777-210 101777-212
2-Phenylpropionitrile ≥97%
2-Phenylpropionitrile

Hazard: Toxic/Lachrymator

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 230-232°C
Density: 0.985
Flash Pt: 93°C
MDL Number: MFCD00001871
CAS Number: 1823-91-2

Order Now

Specification Test Results

Density 0.985
ND20 1.5106

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR