4-Fluorophenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-4-isothiocyanatobenzene

003879-25G 003879-100G
101777-208EA 515.38 USD
101777-208 102550-832
4-Fluorophenyl isothiocyanate ≥98%
4-Fluorophenyl isothiocyanate

TCA listed: T
Hazard: Toxic/Corrosive/Refrigerate/Lachrymator

Formula: C₇H₄FNS
MW: 153.18 g/mol
Boiling Pt: 228 °C
Melting Pt: 26…28 °C
MDL Number: MFCD00004809
CAS Number: 1544-68-9
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Boiling point 228°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR