2-Ethoxyphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Ethoxyphenylisothiocyanate

003878-5G 003878-25G 003878-1G
101777-202EA 100 USD
101777-202 101777-204 101777-200
2-Ethoxyphenyl isothiocyanate ≥98%
2-Ethoxyphenyl isothiocyanate

Hazard: Toxic/Refrigerate/Lachrymator

Formula: C₉H₉NOS
MW: 179.24 g/mol
Boiling Pt: 223-225°C
Density: 1.14
MDL Number: MFCD00046793
CAS Number: 23163-84-0

Order Now

Specification Test Results

Boiling point 223-225°
Density 1.14

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR