3-Fluorophenyl isothiocyanate ≥99%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-3-isothiocyanatobenzene

003876-5G 003876-25G
101777-196EA 70.77 USD
101777-196 101777-198
3-Fluorophenyl isothiocyanate ≥99%
3-Fluorophenyl isothiocyanate

TCA listed: T
Hazard: Toxic/Corrosive/Lachrymator

Formula: C₇H₄FNS
MW: 153.18 g/mol
Boiling Pt: 130°C/30mm
Density: 1.27
MDL Number: MFCD00004804
CAS Number: 404-72-8
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Boiling point 130°/30mm
Density 1.27
ND20 1.62

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR