3-Methylphenethylamine ≥98%

Supplier: Matrix Scientific

003855-1G 003855-5G
101777-178EA 64.62 USD
101777-178 101777-180
3-Methylphenethylamine ≥98%
3-Methylphenethylamine

Hazard: Corrosive

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 110°C/20mm
Density: 0.94
MDL Number: MFCD01310828
CAS Number: 55755-17-4

Order Now

Specification Test Results

Boiling point 110°/20mm
Density 0.94

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR