2-Methylphenethylamine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(o-Tolyl)ethylamine

003854-1G 003854-5G
101777-174EA 60 USD
101777-174 101777-176
2-Methylphenethylamine ≥98%
2-Methylphenethylamine

Hazard: Corrosive

Formula: C₉H₁₃N
Boiling Pt: 97 °C (5 mmHg)
Density: 0.96
MDL Number: MFCD01310827
CAS Number: 55755-16-3

Order Now

Specification Test Results

Boiling point 97°/5mm
Density 0.96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR