Ethyl trifluoropyruvate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate
003819-25G
003819-100G
101777-158EA
26.15
USD
101777-158
101777-160
Ethyl trifluoropyruvate ≥97%
Ethyl trifluoropyruvate
Hazard: Flammable/Refrigerate
Formula:
C₅H₅F₃O₃ MW: 170.09 Boiling Pt: 95…98 °C Density: 1.4 Flash Pt: 27 °C |
MDL Number:
MFCD00114935 CAS Number: 13081-18-0 |
Specification Test Results
Boiling point | 95-98° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...