3,3,3-Trifluoropropionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3,3,3-Trifluoropropanoic acid

003816-25G 003816-5G
101777-154EA 126.15 USD
101777-154 101777-152
3,3,3-Trifluoropropionic acid ≥98%
3,3,3-Trifluoropropionic acid

Hazard: Corrosive

Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 146°C
Melting Pt: 0°C
Density: 1.441
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 146°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR