3,3,3-Trifluoro-1-propanol ≥97%

Supplier: Matrix Scientific

Synonyms: 3,3,3-Trifluoropropan-1-ol

003815-25G 003815-5G
101777-150EA 284.62 USD
101777-150 101777-148
3,3,3-Trifluoro-1-propanol ≥97%
3,3,3-Trifluoro-1-propanol

Hazard: Flammable

Formula: C₃H₅F₃O
Boiling Pt: 100-101°C
Density: 1.294
MDL Number: MFCD00190649
CAS Number: 2240-88-2
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Boiling point 100-101°
Density 1.294
ND20 1.32


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR