(3,4,5-Trifluorophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(3,4,5-Trifluorophenyl)acetonitrile, 3,4,5-Trifluorobenzyl cyanide

003796-5G 003796-1G
101777-136EA 509.23 USD
101777-136 101777-134
(3,4,5-Trifluorophenyl)acetonitrile ≥97%
(3,4,5-Trifluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₄F₃N
MW: 171.12 g/mol
Melting Pt: 30-32°C
MDL Number: MFCD00083531
CAS Number: 220228-03-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR