(2,4,6-Trifluorophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Trifluorobenzyl cyanide, 2-(2,4,6-Trifluorophenyl)acetonitrile

003795-5G 003795-1G
101777-132EA 215.38 USD
101777-132 101777-130
(2,4,6-Trifluorophenyl)acetonitrile ≥97%
(2,4,6-Trifluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₄F₃N
MW: 171.12 g/mol
MDL Number: MFCD00061221
CAS Number: 220227-80-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR