(2,3,6-Trifluorophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(2,3,6-Trifluorophenyl)acetonitrile, 2,3,6-Trifluorobenzyl cyanide

003793-1G 003793-5G
101777-122EA 60 USD
101777-122 101777-124
(2,3,6-Trifluorophenyl)acetonitrile ≥97%
(2,3,6-Trifluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₄F₃N
MW: 171.12 g/mol
Boiling Pt: 80-85°C/5 Torr
MDL Number: MFCD00061219
CAS Number: 114152-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR