(2,3,5-Trifluorophenyl)acetonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
2,3,5-Trifluorobenzyl cyanide, 2,3,5-Trifluorophenylacetonitrile
003792-1G
003792-5G
101777-118EA
141.54
USD
101777-118
101777-120
(2,3,5-Trifluorophenyl)acetonitrile ≥97%
(2,3,5-Trifluorophenyl)acetonitrile
Hazard: Toxic
Formula:
C₈H₄F₃N MW: 171.12 g/mol |
MDL Number:
MFCD00083546 CAS Number: 243666-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...