(2,3,4-Trifluorophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(2,3,4-Trifluorophenyl)acetonitrile, 2,3,4-Trifluorobenzyl cyanide

003791-1G 003791-5G
101777-114EA 81.54 USD
101777-114 101777-116
(2,3,4-Trifluorophenyl)acetonitrile ≥97%
(2,3,4-Trifluorophenyl)acetonitrile

Hazard: Toxic/Moisture Sensitive

Formula: C₈H₄F₃N
MW: 171.12 g/mol
MDL Number: MFCD00061234
CAS Number: 243666-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR