2,2,2-Trifluoroethyl trifluoromethanesulfonate ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2,2-Trifluoroethyltrifluoromethanesulphonate
003720-5G
003720-100G
003720-25G
101777-070EA
20
USD
101777-070
101777-074
101777-072
2,2,2-Trifluoroethyl trifluoromethanesulfonate ≥97%
2,2,2-Trifluoroethyl trifluoromethanesulphonate
Hazard: Toxic/Corrosive
Formula:
C₃H₂F₆O₃S MW: 232.10 g/mol Boiling Pt: 89-91°C/740mm Density: 1.611 |
MDL Number:
MFCD00671579 CAS Number: 6226-25-1 |
Specification Test Results
Boiling point | 89-91°/740 |
ND20 | 1.3053 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...