2,2,2-Trifluoroethyl acetate ≥97%

Supplier: Matrix Scientific

Synonyms: Acetic acid (2,2,2-trifluoroethyl) ester

003718-100G 003718-25G
101777-068EA 330.77 USD
101777-068 101777-066
2,2,2-Trifluoroethyl acetate ≥97%
2,2,2-Trifluoroethyl acetate

TCA listed: T
Hazard: Flammable

Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
Boiling Pt: 78°C
MDL Number: MFCD00198018
CAS Number: 406-95-1
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Boiling point 78°
ND20 1.32

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR