2,2,2-Trifluoroethyl methacrylate ≥99%

Supplier: Matrix Scientific

Synonyms: Methacrylic acid 2,2,2-trifluoroethyl ester

003716-100G 003716-500G
101777-062EA 100 USD
101777-062 101777-064
2,2,2-Trifluoroethyl methacrylate ≥99%
2,2,2-Trifluoroethyl methacrylate

TCA listed: T
Hazard: Flammable/Irritant

Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 59 °C (100 mmHg)
Density: 1.181
MDL Number: MFCD00013576
CAS Number: 352-87-4
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Boiling point 59°/100mm
Density 1.181
ND20 1.361


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR