2,2,2-Trifluoroethyl methyl ether ≥97%

Supplier: Matrix Scientific

003710-25G 003710-5G
101777-050EA 198.46 USD
101777-050 101777-048
2,2,2-Trifluoroethyl methyl ether ≥97%
2,2,2-Trifluoroethyl methyl ether

Hazard: Flammable/Refrigerate

Formula: C₃H₅F₃O
MW: 114.07 g/mol
Boiling Pt: 30°C
Density: 1.576
MDL Number: MFCD00236114
CAS Number: 460-43-5
UN: 1993
ADR: 3,I

Order Now

Specification Test Results

Boiling point 30°
Density 1.576

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR